Assay Details
Target Gene Details
Entrez Gene ID: | 100289388 |
Gene Name: | KCTD21 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:78139793-78173977 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD21-AS1 | NR_102280.1 | |||
| NR_102281.1 | ||||
| DA156609.1 | ||||
| DA737651.1 | ||||
| DA814941.1 | ||||
| DB082989.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
|
| nsv516503 | Chr.11:78145131 - 78209884 on Build GRCh38 | Loss |
|

Genomic Map