Assay Details
Target Gene Details
Entrez Gene ID: | 65084 |
Gene Name: | transmembrane protein 135 |
Gene Aliases: |
PMP52 |
Location: |
Chr.11:87037844-87328834 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM135 | NM_001168724.1 | NP_001162195.1 | ||
| NM_022918.3 | NP_075069.3 | |||
| NR_033149.1 | ||||
| XM_017018140.1 | XP_016873629.1 | |||
| XM_017018141.1 | XP_016873630.1 | |||
| AK000684.1 | ||||
| AK025757.1 | BAB15233.1 | |||
| AK225364.1 | ||||
| AK291804.1 | ||||
| AK296774.1 | ||||
| AK302473.1 | ||||
| BC030952.1 | AAH30952.1 | |||
| BC051462.1 | AAH51462.2 | |||
| BX648678.1 | CAH10546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627114 | Chr.11:87068041 - 87192374 on Build GRCh38 | Gain |
|
| nsv555706 | Chr.11:87077674 - 87115380 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map