Assay Details
Target Gene Details
Entrez Gene ID: | 89797 |
Gene Name: | neuron navigator 2 |
Gene Aliases: |
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2 |
Location: |
Chr.11:19345200-20121601 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAV2 | NM_001111018.1 | NP_001104488.1 | ||
| XM_011520452.2 | XP_011518754.1 | |||
| XM_017018520.1 | XP_016874009.1 | |||
| XM_017018522.1 | XP_016874011.1 | |||
| AB063115.1 | BAC00853.1 |
Target Gene Details
Entrez Gene ID: | 100874012 |
Gene Name: | NAV2 antisense RNA 5 |
Gene Aliases: |
- |
Location: |
Chr.11:19502672-19507229 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAV2-AS5 | NR_049725.1 |
More Information
Set Membership: |
|

Genomic Map