Assay Details
Target Gene Details
Entrez Gene ID: | 23705 |
Gene Name: | cell adhesion molecule 1 |
Gene Aliases: |
BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1 |
Location: |
Chr.11:115173625-115504523 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CADM1 | NM_001098517.1 | NP_001091987.1 | ||
| NM_001301043.1 | NP_001287972.1 | |||
| NM_001301044.1 | NP_001287973.1 | |||
| NM_001301045.1 | NP_001287974.1 | |||
| NM_014333.3 | NP_055148.3 | |||
| XM_005271494.2 | XP_005271551.1 | |||
| XM_017017459.1 | XP_016872948.1 | |||
| XM_017017460.1 | XP_016872949.1 | |||
| XM_017017461.1 | XP_016872950.1 | |||
| AB094146.1 | BAC66178.1 | |||
| AF132811.1 | AAF69029.1 | |||
| AK075502.1 | BAC11657.1 | |||
| AK315245.1 | ||||
| AY358334.1 | AAQ88700.1 | |||
| BC047021.1 | AAH47021.1 | |||
| BC125102.1 | ||||
| BC125103.1 | ||||
| BI915307.1 | ||||
| CR457157.1 | CAG33438.1 | |||
| HE586496.1 | ||||
| HE586497.1 | ||||
| HE586498.1 | ||||
| HE586499.1 | ||||
| HE586500.1 | ||||
| HE586503.1 | ||||
| KJ534791.1 | ||||
| KJ534792.1 | ||||
| KJ534793.1 | ||||
| KJ534794.1 | ||||
| KJ534795.1 | ||||
| KJ534796.1 | ||||
| KJ534797.1 |
Target Gene Details
Entrez Gene ID: | 105369509 |
Gene Name: | uncharacterized LOC105369509 |
Gene Aliases: |
- |
Location: |
Chr.11:115333577-115340439 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105369509 | NR_135108.1 | |||
| BX119732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042473 | Chr.11:115334028 - 115400481 on Build GRCh38 | Loss |
|
| esv3627810 | Chr.11:115295545 - 115341460 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map