Assay Details
Target Gene Details
Entrez Gene ID: | 50863 |
Gene Name: | neurotrimin |
Gene Aliases: |
HNT, IGLON2, NTRI |
Location: |
Chr.11:131370326-132336822 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTM | NM_001048209.1 | NP_001041674.1 | ||
| XM_005271580.3 | XP_005271637.1 | |||
| XM_005271582.3 | XP_005271639.1 | |||
| XM_006718855.3 | XP_006718918.1 | |||
| XM_017017852.1 | XP_016873341.1 | |||
| XM_017017853.1 | XP_016873342.1 | |||
| XM_017017855.1 | XP_016873344.1 | |||
| XM_017017861.1 | XP_016873350.1 | |||
| XM_017017865.1 | XP_016873354.1 | |||
| AK293487.1 | ||||
| AK294137.1 | ||||
| AK315964.1 | ||||
| AY358331.1 | AAQ88697.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892109 | Chr.11:131358314 - 131494601 on Build GRCh38 | Loss |
|
| nsv1035733 | Chr.11:130509478 - 131429821 on Build GRCh38 | Gain |
|
| dgv1297n100 | Chr.11:131036593 - 131374908 on Build GRCh38 | Gain |
|
| nsv1039087 | Chr.11:131230668 - 131438530 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map