Assay Details
Target Gene Details
Entrez Gene ID: | 5612 |
Gene Name: | THAP domain containing 12 |
Gene Aliases: |
DAP4, P52rIPK, PRKRIR, THAP0 |
Location: |
Chr.11:76349956-76380965 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THAP12 | NM_004705.3 | NP_004696.2 | ||
| NR_130898.1 | ||||
| AF007393.1 | AAC39564.1 | |||
| AF081567.1 | AAG01570.1 | |||
| AK291843.1 | ||||
| BC110304.1 | AAI10305.1 | |||
| BC117138.1 | ||||
| BC117140.1 | ||||
| DR005363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521451 | Chr.11:76342483 - 76376800 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map