Assay Details
Target Gene Details
Entrez Gene ID: | 54851 |
Gene Name: | ankyrin repeat domain 49 |
Gene Aliases: |
FGIF, GBIF |
Location: |
Chr.11:94493987-94499578 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD49 | NM_017704.2 | NP_060174.2 | ||
| XM_017017941.1 | XP_016873430.1 | |||
| AF025354.1 | AAK60412.1 | |||
| AK000196.1 | BAA91003.1 | |||
| AK000448.1 | ||||
| AL833977.1 | CAD38821.2 | |||
| BC017798.1 | AAH17798.1 | |||
| BG025192.1 |
Target Gene Details
Entrez Gene ID: | 4361 |
Gene Name: | MRE11 homolog A, double strand break repair nuclease |
Gene Aliases: |
ATLD, HNGS1, MRE11, MRE11B |
Location: |
Chr.11:94415570-94512701 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRE11A | XM_011542837.2 | XP_011541139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2671948 | Chr.11:94349534 - 94645424 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map