Assay Details
Target Gene Details
Entrez Gene ID: | 51296 |
Gene Name: | solute carrier family 15 member 3 |
Gene Aliases: |
OCTP, PHT2, PTR3 |
Location: |
Chr.11:60937083-60952081 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC15A3 | NM_016582.2 | NP_057666.1 | ||
| NR_027391.1 | ||||
| XM_011545095.2 | XP_011543397.1 | |||
| AB020598.1 | BAA93432.1 | |||
| AF135600.1 | AAQ13565.1 | |||
| AK127216.1 | 1 | 3020 | ||
| BC025710.1 | ||||
| BC037974.1 | AAH37974.1 |
More Information
Set Membership: |
|

Genomic Map