Assay Details
Target Gene Details
Entrez Gene ID: | 747 |
Gene Name: | diacylglycerol lipase alpha |
Gene Aliases: |
C11orf11, DAGL(ALPHA), DAGLALPHA, NSDDR |
Location: |
Chr.11:61680184-61747002 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAGLA | NM_006133.2 | NP_006124.1 | ||
| XM_017018238.1 | XP_016873727.1 | |||
| XM_017018240.1 | XP_016873729.1 | |||
| AB014559.2 | ||||
| BC150176.1 | ||||
| BC150195.1 | ||||
| BC152453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832179 | Chr.11:61605799 - 61757528 on Build GRCh38 | Loss |
|
| esv1003572 | Chr.11:61677768 - 61689740 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map