Assay Details
Target Gene Details
Entrez Gene ID: | 1408 |
Gene Name: | cryptochrome circadian clock 2 |
Gene Aliases: |
HCRY2, PHLL2 |
Location: |
Chr.11:45847118-45883248 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRY2 | NM_001127457.2 | NP_001120929.1 | ||
| NM_021117.3 | NP_066940.2 | |||
| AB014558.1 | BAA31633.1 | |||
| AK291260.1 | ||||
| AK294904.1 | ||||
| AK302865.1 | ||||
| BC035161.1 | AAH35161.1 | |||
| BC041814.1 | AAH41814.1 | |||
| DC330290.1 | ||||
| EU219622.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs80354836] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map