Assay Details
Target Gene Details
Entrez Gene ID: | 120103 |
Gene Name: | solute carrier family 36 member 4 |
Gene Aliases: |
PAT4 |
Location: |
Chr.11:93144171-93197983 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC36A4 | NM_001286139.1 | 11 | 2731 | NP_001273068.1 |
| NM_152313.3 | 11 | 2694 | NP_689526.2 | |
| XM_017017174.1 | 11 | 2685 | XP_016872663.1 | |
| XM_017017175.1 | 10 | 2611 | XP_016872664.1 | |
| XM_017017176.1 | 10 | 2563 | XP_016872665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039134 | Chr.11:93056834 - 93486776 on Build GRCh38 | Loss |
|
| nsv435 | Chr.11:93120600 - 93154727 on Build GRCh38 | Deletion |
|
| dgv2094n54 | Chr.11:92907621 - 93148767 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map