Assay Details
Target Gene Details
Entrez Gene ID: | 105376671 |
Gene Name: | uncharacterized LOC105376671 |
Gene Aliases: |
lnc-BBOX1-1 |
Location: |
Chr.11:27471731-27482433 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105376671 | NR_131169.1 | |||
| NR_131170.1 | ||||
| AF150379.1 | ||||
| DA085906.1 |

Genomic Map