Assay Details
Target Gene Details
Entrez Gene ID: | 643 |
Gene Name: | C-X-C motif chemokine receptor 5 |
Gene Aliases: |
BLR1, CD185, MDR15 |
Location: |
Chr.11:118883766-118896271 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCR5 | NM_001716.4 | NP_001707.1 | ||
| AK291512.1 | ||||
| BC110352.1 | AAI10353.1 | |||
| DA815880.1 | ||||
| X68149.1 | CAA48252.1 |
More Information
Set Membership: |
|

Genomic Map