Assay Details
Target Gene Details
Entrez Gene ID: | 105369535 |
Gene Name: | uncharacterized LOC105369535 |
Gene Aliases: |
- |
Location: |
Chr.11:121472920-121484806 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105369535 | XM_017018646.1 | XP_016874135.1 | ||
| XM_017018647.1 | XP_016874136.1 | |||
| XM_017018648.1 | XP_016874137.1 | |||
| XM_017018649.1 | XP_016874138.1 | |||
| XM_017018650.1 | XP_016874139.1 | |||
| XM_017018651.1 | XP_016874140.1 |
Target Gene Details
Entrez Gene ID: | 6653 |
Gene Name: | sortilin related receptor 1 |
Gene Aliases: |
C11orf32, LR11, LRP9, SORLA, SorLA-1, gp250 |
Location: |
Chr.11:121452203-121633762 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SORL1 | NM_003105.5 | NP_003096.1 | ||
| XM_011542963.2 | XP_011541265.1 | |||
| XM_017018169.1 | XP_016873658.1 | |||
| XM_017018171.1 | XP_016873660.1 | |||
| AK309523.1 | ||||
| BC137171.1 | ||||
| BX365721.2 | ||||
| DN831521.1 | ||||
| Y08110.1 | CAA69325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv24898 | Chr.11:121480508 - 121481978 on Build GRCh38 | Loss |
|
| nsv1141204 | Chr.11:121480291 - 121481991 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map