Assay Details
Target Gene Details
Entrez Gene ID: | 54970 |
Gene Name: | tetratricopeptide repeat domain 12 |
Gene Aliases: |
TPARM |
Location: |
Chr.11:113314529-113385952 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC12 | NM_001318533.1 | NP_001305462.1 | ||
| NM_017868.3 | NP_060338.3 | |||
| XM_005271604.3 | XP_005271661.1 | |||
| XM_011542890.2 | XP_011541192.1 | |||
| XM_017017943.1 | XP_016873432.1 | |||
| XM_017017944.1 | XP_016873433.1 | |||
| XM_017017945.1 | XP_016873434.1 | |||
| XM_017017946.1 | XP_016873435.1 | |||
| XM_017017947.1 | XP_016873436.1 | |||
| XM_017017948.1 | XP_016873437.1 | |||
| XM_017017949.1 | XP_016873438.1 | |||
| XM_017017950.1 | XP_016873439.1 | |||
| AK000542.1 | BAA91242.1 | |||
| AK023921.1 | BAB14725.1 | |||
| AK091887.1 | ||||
| AK223117.1 | BAD96837.1 | |||
| AK292331.1 | ||||
| DA954030.1 | ||||
| DB110945.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483080 | Chr.11:113216072 - 113400474 on Build GRCh38 | Loss |
|

Genomic Map