Assay Details
Target Gene Details
Entrez Gene ID: | 80168 |
Gene Name: | monoacylglycerol O-acyltransferase 2 |
Gene Aliases: |
DGAT2L5, DGAT2L5., MGAT2, hDC5 |
Location: |
Chr.11:75717819-75732958 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOGAT2 | NM_025098.3 | NP_079374.2 | ||
| XM_011545267.1 | XP_011543569.1 | |||
| AK026297.1 | BAB15436.1 | |||
| AK291998.1 | ||||
| AY157608.1 | AAO23672.1 | |||
| BC103876.1 | AAI03877.1 | |||
| BC103877.1 | AAI03878.1 | |||
| BC103878.1 | AAI03879.1 | |||
| DA922421.1 |
More Information
Set Membership: |
|

Genomic Map