Assay Details
Target Gene Details
Entrez Gene ID: | 54948 |
Gene Name: | mitochondrial ribosomal protein L16 |
Gene Aliases: |
L16mt, MRP-L16, PNAS-111 |
Location: |
Chr.11:59806135-59810872 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL16 | NM_017840.3 | NP_060310.1 | ||
| AB049642.1 | BAB40847.1 | |||
| AF183428.1 | AAG09697.1 | |||
| AK000491.1 | BAA91202.1 | |||
| BC001040.2 | AAH01040.1 | |||
| BC019269.1 | AAH19269.1 | |||
| CR457117.1 | CAG33398.1 |
More Information
Set Membership: |
|

Genomic Map