Assay Details
Target Gene Details
Entrez Gene ID: | 160335 |
Gene Name: | transmembrane and tetratricopeptide repeat containing 2 |
Gene Aliases: |
IBDBP1 |
Location: |
Chr.12:82686952-83134868 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMTC2 | NM_001320321.1 | NP_001307250.1 | ||
| NM_001320322.1 | NP_001307251.1 | |||
| NM_152588.2 | NP_689801.1 | |||
| XM_017018884.1 | XP_016874373.1 | |||
| AK299771.1 | ||||
| AK315285.1 | ||||
| AL834509.1 | CAD39165.1 | |||
| AX795392.1 | CAE48265.1 | |||
| BC093852.1 | AAH93852.1 | |||
| BC093854.1 | AAH93854.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521849 | Chr.12:82620004 - 83597700 on Build GRCh38 | Gain |
|
| nsv469490 | Chr.12:82695652 - 83113490 on Build GRCh38 | Loss |
|
| nsv559541 | Chr.12:82321431 - 82714616 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map