Assay Details
Target Gene Details
Entrez Gene ID: | 10094 |
Gene Name: | actin related protein 2/3 complex subunit 3 |
Gene Aliases: |
ARC21, p21-Arc |
Location: |
Chr.12:110434890-110450411 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPC3 | NM_001278556.1 | NP_001265485.1 | ||
| NM_001287222.1 | NP_001274151.1 | |||
| AF004561.1 | AAB61466.1 | |||
| AF006086.1 | AAB64191.1 | |||
| AK297309.1 | ||||
| AK311789.1 | ||||
| AL357196.1 | ||||
| BC067747.1 | AAH67747.1 | |||
| BC078162.1 | AAH78162.1 | |||
| BG620669.1 | ||||
| BG723870.1 | ||||
| CR407667.1 | CAG28595.1 | |||
| DQ328220.1 | ABC59627.1 |
More Information
Set Membership: |
|

Genomic Map