Assay Details
Target Gene Details
Entrez Gene ID: | 83758 |
Gene Name: | retinol binding protein 5 |
Gene Aliases: |
CRBP-III, CRBP3, CRBPIII, HRBPiso |
Location: |
Chr.12:7119106-7130272 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBP5 | XM_017020002.1 | XP_016875491.1 | ||
| AF212239.1 | AAK14925.1 | |||
| AK096947.1 | ||||
| AK223378.1 | BAD97098.1 | |||
| AK311977.1 | ||||
| AY007436.1 | AAG09617.1 | |||
| BC029355.1 | AAH29355.1 |
More Information
Set Membership: |
|

Genomic Map