Assay Details
Target Gene Details
Entrez Gene ID: | 8848 |
Gene Name: | TSC22 domain family member 1 |
Gene Aliases: |
Ptg-2, TGFB1I4, TSC22 |
Location: |
Chr.13:44432143-44576565 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D1 | NM_001243798.1 | NP_001230727.1 | ||
| NM_001243799.1 | NP_001230728.1 | |||
| NM_183422.3 | NP_904358.2 | |||
| AB082525.1 | ||||
| AK027071.1 | ||||
| AK091854.1 | ||||
| BC136475.1 | ||||
| BC136482.1 | ||||
| BI463370.1 | ||||
| CR627459.1 | CAH10539.1 |
More Information
Set Membership: |
|

Genomic Map