Assay Details
Target Gene Details
Entrez Gene ID: | 57096 |
Gene Name: | retinitis pigmentosa GTPase regulator interacting protein 1 |
Gene Aliases: |
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d |
Location: |
Chr.14:21287977-21351316 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPGRIP1 | NM_020366.3 | NP_065099.3 | ||
| AJ417067.1 | CAD01135.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054778 | Chr.14:21294781 - 21334420 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map