Assay Details
Target Gene Details
Entrez Gene ID: | 3183 |
Gene Name: | heterogeneous nuclear ribonucleoprotein C (C1/C2) |
Gene Aliases: |
C1, C2, HNRNP, HNRPC, SNRPC |
Location: |
Chr.14:21209136-21269479 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPC | NM_001077442.1 | 8 | 2739 | NP_001070910.1 |
| NM_001077443.1 | 8 | 2700 | NP_001070911.1 | |
| NM_004500.3 | 9 | 2726 | NP_004491.2 | |
| NM_031314.2 | 9 | 2765 | NP_112604.2 | |
| AL833432.1 | 6 | 2303 |
More Information
Set Membership: |
|

Genomic Map