Assay Details
Target Gene Details
Entrez Gene ID: | 9950 |
Gene Name: | golgin A5 |
Gene Aliases: |
GOLIM5, RFG5, ret-II |
Location: |
Chr.14:92794231-92839963 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLGA5 | NM_005113.3 | NP_005104.3 | ||
| XM_011537420.2 | XP_011535722.1 | |||
| AF085199.1 | AAD09753.1 | |||
| AJ132949.1 | CAB36967.1 | |||
| AK293077.1 | ||||
| AK303789.1 | ||||
| AY644768.1 | AAV85456.1 | |||
| BC023021.1 | AAH23021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522417 | Chr.14:92811952 - 92824114 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map