Assay Details
Target Gene Details
Entrez Gene ID: | 10588 |
Gene Name: | 5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase) |
Gene Aliases: |
HsT19268 |
Location: |
Chr.15:79843547-79897285 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTHFS | NM_001199758.1 | NP_001186687.1 | ||
| NM_006441.3 | NP_006432.1 | |||
| NR_037654.1 | ||||
| AL110101.1 | ||||
| BC012417.1 | AAH12417.1 | |||
| BC019921.1 | AAH19921.1 | |||
| BM450865.1 | ||||
| CB995322.1 | ||||
| CK905549.1 | ||||
| L38928.1 | AAC41945.1 |
Target Gene Details
Entrez Gene ID: | 100528021 |
Gene Name: | ST20-MTHFS readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:79843547-79923106 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST20-MTHFS | NM_001199760.1 | NP_001186689.1 | ||
| BG705061.1 | ||||
| CK002935.1 | ||||
| DB495490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637008 | Chr.15:79874155 - 79879033 on Build GRCh38 | Loss |
|
| nsv833065 | Chr.15:79760661 - 79982480 on Build GRCh38 | Loss |
|
| esv2678719 | Chr.15:79874161 - 79879033 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs149183525] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map