Assay Details
Target Gene Details
Entrez Gene ID: | 54962 |
Gene Name: | TIMELESS interacting protein |
Gene Aliases: |
- |
Location: |
Chr.15:66334976-66386787 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIPIN | NM_001289986.1 | NP_001276915.1 | ||
| NM_017858.2 | NP_060328.2 | |||
| XM_005254521.2 | XP_005254578.1 | |||
| XM_005254522.4 | XP_005254579.1 | |||
| XM_006720594.3 | XP_006720657.1 | |||
| XM_011521743.1 | XP_011520045.1 | |||
| XM_017022390.1 | XP_016877879.1 | |||
| XM_017022391.1 | XP_016877880.1 | |||
| AK000523.1 | BAA91229.1 | |||
| AK299233.1 | ||||
| BC000870.1 | AAH00870.1 | |||
| BU596053.1 | ||||
| BU688750.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047588 | Chr.15:66227169 - 66394432 on Build GRCh38 | Loss |
|
| nsv833045 | Chr.15:66295248 - 66460419 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map