Assay Details
Target Gene Details
Entrez Gene ID: | 9245 |
Gene Name: | glucosaminyl (N-acetyl) transferase 3, mucin type |
Gene Aliases: |
C2/4GnT, C24GNT, C2GNT2, C2GNTM, GNTM |
Location: |
Chr.15:59611783-59620011 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCNT3 | NM_004751.2 | NP_004742.1 | ||
| BC017032.1 | AAH17032.1 | |||
| BP243086.1 | ||||
| EF152283.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain |
|
| esv3636644 | Chr.15:59550375 - 59641686 on Build GRCh38 | Loss |
|
| esv2668729 | Chr.15:59550283 - 59641656 on Build GRCh38 | Deletion |
|
| nsv569605 | Chr.15:59563027 - 59664354 on Build GRCh38 | Loss |
|
| esv23994 | Chr.15:59595995 - 59612450 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78952803] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map