Assay Details
Target Gene Details
Entrez Gene ID: | 91860 |
Gene Name: | calmodulin like 4 |
Gene Aliases: |
NY-BR-20 |
Location: |
Chr.15:68190705-68206110 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALML4 | NM_001031733.2 | NP_001026903.2 | ||
| NM_001286694.1 | NP_001273623.1 | |||
| NM_001286695.1 | NP_001273624.1 | |||
| NM_033429.2 | NP_219501.2 | |||
| NR_104583.1 | ||||
| AF308287.1 | AAG48255.1 | |||
| AK092628.1 | ||||
| AK296804.1 | ||||
| BX640702.1 | CAE45822.1 | |||
| BX640817.1 | CAE45894.1 | |||
| DB204573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1581 | Chr.15:68172970 - 68204223 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map