Assay Details
Target Gene Details
Entrez Gene ID: | 89978 |
Gene Name: | diphthamine biosynthesis 6 |
Gene Aliases: |
ATPBD4 |
Location: |
Chr.15:35217356-35546222 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPH6 | XM_017022708.1 | XP_016878197.1 | ||
| BX537862.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3552302 | Chr.15:35237800 - 35243799 on Build GRCh38 | Deletion |
|
| nsv1486 | Chr.15:35202047 - 35244348 on Build GRCh38 | Deletion |
|
| esv2464651 | Chr.15:35238381 - 35244268 on Build GRCh38 | Deletion |
|
| nsv1487 | Chr.15:35228589 - 35258344 on Build GRCh38 | Insertion |
|
| nsv952609 | Chr.15:35236600 - 35241999 on Build GRCh38 | Deletion |
|
| esv3636132 | Chr.15:35238426 - 35243735 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map