Assay Details
Target Gene Details
Entrez Gene ID: | 51496 |
Gene Name: | CTD small phosphatase like 2 |
Gene Aliases: |
HSPC058, HSPC129 |
Location: |
Chr.15:44427329-44527774 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTDSPL2 | NM_016396.2 | NP_057480.2 | ||
| XM_005254441.2 | XP_005254498.1 | |||
| XM_006720565.2 | XP_006720628.1 | |||
| XM_011521665.2 | XP_011519967.1 | |||
| XM_017022306.1 | XP_016877795.1 | |||
| XM_017022307.1 | XP_016877796.1 | |||
| AF161478.1 | AAF29093.1 | |||
| AF161543.1 | AAF29030.1 | |||
| AK226094.1 | ||||
| AK291230.1 | ||||
| BC018623.1 | ||||
| BC035744.1 | AAH35744.1 | |||
| DA995235.1 | ||||
| DQ128104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4510n54 | Chr.15:44306444 - 44483877 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map