Assay Details
Target Gene Details
Entrez Gene ID: | 197021 |
Gene Name: | lactase like |
Gene Aliases: |
KLG, KLPH |
Location: |
Chr.15:66547468-66566025 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCTL | NM_001278562.1 | NP_001265491.1 | ||
| NR_103739.1 | ||||
| XM_017021999.1 | XP_016877488.1 | |||
| AK090598.1 | ||||
| AK311441.1 |
More Information
Set Membership: |
|

Genomic Map