Assay Details
Target Gene Details
Entrez Gene ID: | 60490 |
Gene Name: | phosphopantothenoylcysteine decarboxylase |
Gene Aliases: |
MDS018, PPC-DC, coaC |
Location: |
Chr.15:75023533-75060180 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPCDC | NM_001301101.1 | NP_001288030.1 | ||
| NM_001301102.1 | NP_001288031.1 | |||
| NM_001301103.1 | NP_001288032.1 | |||
| NM_001301104.1 | NP_001288033.1 | |||
| NM_001301105.1 | NP_001288034.1 | |||
| NM_021823.4 | NP_068595.3 | |||
| NR_125369.1 | ||||
| XM_017022461.1 | XP_016877950.1 | |||
| XM_017022462.1 | XP_016877951.1 | |||
| XM_017022463.1 | 1 | 1329 | XP_016877952.1 | |
| XM_017022464.1 | XP_016877953.1 | |||
| XM_017022465.1 | XP_016877954.1 | |||
| AF182419.1 | AAG14955.1 | |||
| AK027491.1 | BAB55151.1 | |||
| AY358848.1 | AAQ89207.1 | |||
| BC014409.1 | AAH14409.1 | |||
| BF243417.1 | ||||
| BG744258.1 | ||||
| BI833814.1 | ||||
| BI907452.1 | ||||
| BU500301.1 | ||||
| DB488540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569994 | Chr.15:75044388 - 75155432 on Build GRCh38 | Loss |
|
| nsv569993 | Chr.15:74949814 - 75191411 on Build GRCh38 | Gain |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map