Assay Details
Target Gene Details
Entrez Gene ID: | 90525 |
Gene Name: | Src homology 2 domain containing F |
Gene Aliases: |
- |
Location: |
Chr.15:45167214-45201175 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHF | NM_001301168.1 | NP_001288097.1 | ||
| NM_001301169.1 | NP_001288098.1 | |||
| NM_001301170.1 | NP_001288099.1 | |||
| NM_001301171.1 | NP_001288100.1 | |||
| NM_138356.2 | NP_612365.2 | |||
| XM_006720742.3 | XP_006720805.2 | |||
| XM_011522174.2 | XP_011520476.1 | |||
| XM_011522175.2 | XP_011520477.1 | |||
| XM_011522176.2 | XP_011520478.1 | |||
| XM_011522177.2 | XP_011520479.1 | |||
| XM_011522178.2 | XP_011520480.1 | |||
| XM_017022710.1 | XP_016878199.1 | |||
| XM_017022711.1 | XP_016878200.1 | |||
| AK094391.1 | BAC04345.1 | |||
| AK096267.1 | ||||
| AK301625.1 | ||||
| AK303749.1 | ||||
| BX332806.2 | ||||
| BX446025.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581685 | Chr.15:45179512 - 45195934 on Build GRCh38 | Loss |
|
| dgv738e212 | Chr.15:45179245 - 45190509 on Build GRCh38 | Loss |
|
| nsv569315 | Chr.15:45153958 - 45480250 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map