Assay Details
Target Gene Details
Entrez Gene ID: | 161823 |
Gene Name: | adenosine deaminase like |
Gene Aliases: |
- |
Location: |
Chr.15:43330351-43354555 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAL | NM_001159280.2 | 12 | 2763 | NP_001152752.1 |
| NM_001324366.1 | 13 | 2844 | NP_001311295.1 | |
| XM_011521291.2 | 12 | 2320 | XP_011519593.1 | |
| XM_017021964.1 | 13 | 2430 | XP_016877453.1 | |
| XM_017021965.1 | 13 | 2409 | XP_016877454.1 | |
| XM_017021966.1 | 12 | 2346 | XP_016877455.1 | |
| XM_017021967.1 | 11 | 2094 | XP_016877456.1 | |
| XM_017021969.1 | 10 | 2138 | XP_016877458.1 | |
| AK126583.1 | 7 | 1679 | ||
| BX647599.1 | 3 | 2101 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1512 | Chr.15:43324636 - 43369448 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map