Assay Details
Target Gene Details
Entrez Gene ID: | 8800 |
Gene Name: | peroxisomal biogenesis factor 11 alpha |
Gene Aliases: |
PEX11-ALPHA, PMP28, hsPEX11p |
Location: |
Chr.15:89681531-89690784 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEX11A | NM_001271572.1 | NP_001258501.1 | ||
| NM_001271573.1 | NP_001258502.1 | |||
| NM_003847.2 | NP_003838.1 | |||
| AB015594.1 | BAA32533.1 | |||
| AF093668.1 | AAC78658.1 | |||
| AK001415.1 | ||||
| AK297442.1 | ||||
| AK300978.1 | ||||
| AK313318.1 | ||||
| BC009697.1 | AAH09697.1 | |||
| BC101164.2 | ||||
| BC114384.1 | AAI14385.1 | |||
| CR542046.1 | CAG46843.1 | |||
| CR542075.1 | CAG46872.1 | |||
| DA773492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv570387 | Chr.15:89677442 - 89692000 on Build GRCh38 | Loss |
|
| esv3637201 | Chr.15:89678659 - 89694358 on Build GRCh38 | Loss |
|
| nsv977756 | Chr.15:89687413 - 89689672 on Build GRCh38 | Duplication |
|
| nsv1046117 | Chr.15:89683716 - 89765410 on Build GRCh38 | Gain |
|
| nsv1660 | Chr.15:89660975 - 89694859 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map