Assay Details
Target Gene Details
Entrez Gene ID: | 176 |
Gene Name: | aggrecan |
Gene Aliases: |
AGC1, AGCAN, CSPG1, CSPGCP, MSK16, SEDK |
Location: |
Chr.15:88803442-88875354 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAN | NM_001135.3 | NP_001126.3 | ||
| NM_013227.3 | NP_037359.3 | |||
| XM_006720419.1 | XP_006720482.1 | |||
| XM_011521313.1 | XP_011519615.1 | |||
| XM_011521314.1 | XP_011519616.1 | |||
| XM_017021985.1 | XP_016877474.1 | |||
| XM_017021986.1 | XP_016877475.1 | |||
| XM_017021987.1 | XP_016877476.1 | |||
| AK290723.1 | ||||
| BC036445.1 | AAH36445.1 | |||
| DA596050.1 | ||||
| M55172.1 | AAA62824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv570385 | Chr.15:88806095 - 88830608 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map