Assay Details
Target Gene Details
Entrez Gene ID: | 23184 |
Gene Name: | mesoderm development candidate 2 |
Gene Aliases: |
BOCA, MESD |
Location: |
Chr.15:80947326-80989878 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MESDC2 | NR_126327.1 | |||
| NR_126328.1 | ||||
| AK301416.1 | ||||
| BC009210.2 | AAH09210.1 | |||
| BC012746.2 | AAH12746.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115295813] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map