Assay Details
Target Gene Details
Entrez Gene ID: | 645212 |
Gene Name: | EIF3J antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.15:44534505-44536923 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3J-AS1 | NR_034170.1 | 2 | 1431 | |
| NR_034171.1 | 3 | 966 | ||
| AK092547.1 | 2 | 1182 |
More Information
Additional Information:
For this assay, SNP(s) [rs76263379] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map