Assay Details
Target Gene Details
Entrez Gene ID: | 89978 |
Gene Name: | diphthamine biosynthesis 6 |
Gene Aliases: |
ATPBD4 |
Location: |
Chr.15:35217356-35546222 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPH6 | XM_017022708.1 | XP_016878197.1 | ||
| BX537862.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1487 | Chr.15:35228589 - 35258344 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map