Assay Details
Target Gene Details
Entrez Gene ID: | 64799 |
Gene Name: | IQ motif containing H |
Gene Aliases: |
NYDSP5 |
Location: |
Chr.15:67254772-67501804 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Gene Details
Entrez Gene ID: | 100506686 |
Gene Name: | IQCH antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:67403611-67521844 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCH-AS1 | NR_040051.1 | 6 | 1248 | |
| NR_040052.1 | 6 | 1044 |
More Information
Set Membership: |
|

Genomic Map