Assay Details
Target Gene Details
Entrez Gene ID: | 26035 |
Gene Name: | glucuronic acid epimerase |
Gene Aliases: |
HSEPI |
Location: |
Chr.15:69160555-69272209 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLCE | NM_001324091.1 | NP_001311020.1 | ||
| NM_001324092.1 | NP_001311021.1 | |||
| NM_001324093.1 | NP_001311022.1 | |||
| NM_001324094.1 | NP_001311023.1 | |||
| NM_015554.2 | NP_056369.1 | |||
| XM_005254298.3 | XP_005254355.1 | |||
| AY635582.1 | AAT48654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569811 | Chr.15:69215121 - 69225615 on Build GRCh38 | Gain |
|
| nsv1039652 | Chr.15:69204723 - 69258106 on Build GRCh38 | Loss |
|
| nsv517185 | Chr.15:69137677 - 69260783 on Build GRCh38 | Gain+Loss |
|
| esv23604 | Chr.15:69211258 - 69230312 on Build GRCh38 | Gain |
|
| esv3636821 | Chr.15:69211145 - 69230108 on Build GRCh38 | Gain |
|
| dgv4584n54 | Chr.15:69214001 - 69232379 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map