Assay Details
Target Gene Details
Entrez Gene ID: | 64799 |
Gene Name: | IQ motif containing H |
Gene Aliases: |
NYDSP5 |
Location: |
Chr.15:67254772-67501804 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Target Gene Details
Entrez Gene ID: | 100506686 |
Gene Name: | IQCH antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:67403611-67521844 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCH-AS1 | NR_040051.1 | |||
| NR_040052.1 | ||||
| BX332831.2 |

Genomic Map