Assay Details
Target Gene Details
Entrez Gene ID: | 101928635 |
Gene Name: | uncharacterized LOC101928635 |
Gene Aliases: |
- |
Location: |
Chr.15:58106609-58431770 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928635 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1549 | Chr.15:58122615 - 58155766 on Build GRCh38 | Insertion |
|
| nsv1047246 | Chr.15:58083408 - 58414959 on Build GRCh38 | Gain |
|
| nsv569597 | Chr.15:58125696 - 58165308 on Build GRCh38 | Loss |
|

Genomic Map