Assay Details
Target Gene Details
Entrez Gene ID: | 104564224 |
Gene Name: | leukemia-associated non-coding IGF1R activator RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:99014704-99031050 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUNAR1 | NR_126487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055008 | Chr.15:98933915 - 99479656 on Build GRCh38 | Gain |
|
| nsv509583 | Chr.15:98969886 - 99110574 on Build GRCh38 | Insertion |
|
| esv3892741 | Chr.15:98979139 - 99618239 on Build GRCh38 | Gain |
|
| nsv1045124 | Chr.15:98961614 - 99307005 on Build GRCh38 | Gain |
|

Genomic Map