Assay Details
Target Gene Details
Entrez Gene ID: | 55055 |
Gene Name: | zwilch kinetochore protein |
Gene Aliases: |
KNTC1AP, hZwilch |
Location: |
Chr.15:66505083-66549485 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZWILCH | NM_001287821.1 | NP_001274750.1 | ||
| NM_001287822.1 | NP_001274751.1 | |||
| NM_001287823.1 | NP_001274752.1 | |||
| NM_017975.4 | NP_060445.3 | |||
| AK000898.1 | ||||
| AK023175.1 | BAB14446.1 | |||
| AK122787.1 | ||||
| AK291048.1 | ||||
| BC036900.1 | AAH36900.1 | |||
| BC090041.1 | AAH90041.1 | |||
| BX640701.1 | CAE45821.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74894249] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map