Assay Details
Target Gene Details
Entrez Gene ID: | 9915 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator 2 |
Gene Aliases: |
WEDAS, bHLHe1 |
Location: |
Chr.15:80404350-80597937 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARNT2 | NM_014862.3 | NP_055677.3 | ||
| BC036099.1 | AAH36099.1 | |||
| BC051335.1 | AAH51335.1 | |||
| BX538026.1 | CAD97972.1 | |||
| BX647212.1 | ||||
| KJ534772.1 | ||||
| KJ534773.1 | ||||
| KJ534986.1 | ||||
| KJ534995.1 | ||||
| KJ534996.1 | ||||
| KJ535008.1 | ||||
| KJ535009.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78785899] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map