Assay Details
Target Gene Details
Entrez Gene ID: | 366 |
Gene Name: | aquaporin 9 |
Gene Aliases: |
AQP-9, HsT17287, SSC1, T17287 |
Location: |
Chr.15:58138169-58185911 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AQP9 | NM_001320635.1 | NP_001307564.1 | ||
| NM_001320636.1 | NP_001307565.1 | |||
| NM_020980.4 | NP_066190.2 | |||
| AB008775.1 | BAA24864.1 | |||
| AF016495.1 | AAF16677.1 | |||
| AK300055.1 | ||||
| AK313214.1 | ||||
| BC026258.1 | AAH26258.1 | |||
| CR542027.1 | CAG46824.1 | |||
| DC296768.1 | ||||
| DC410284.1 |
Target Gene Details
Entrez Gene ID: | 101928635 |
Gene Name: | uncharacterized LOC101928635 |
Gene Aliases: |
- |
Location: |
Chr.15:58106609-58431770 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928635 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047246 | Chr.15:58083408 - 58414959 on Build GRCh38 | Gain |
|
| nsv569597 | Chr.15:58125696 - 58165308 on Build GRCh38 | Loss |
|
| nsv833024 | Chr.15:58155260 - 58329764 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73426165] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map