Assay Details
Target Gene Details
Entrez Gene ID: | 5597 |
Gene Name: | mitogen-activated protein kinase 6 |
Gene Aliases: |
ERK3, HsT17250, PRKM6, p97MAPK |
Location: |
Chr.15:52004265-52066265 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK6 | XM_005254539.3 | XP_005254596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1536 | Chr.15:51984096 - 52028722 on Build GRCh38 | Deletion |
|
| dgv2595n100 | Chr.15:51964903 - 52049904 on Build GRCh38 | Gain |
|
| nsv471244 | Chr.15:52013266 - 52219268 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map