Assay Details
Target Gene Details
Entrez Gene ID: | 825 |
Gene Name: | calpain 3 |
Gene Aliases: |
CANP3, CANPL3, LGMD2, LGMD2A, nCL-1, p94 |
Location: |
Chr.15:42359500-42412317 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPN3 | NM_000070.2 | NP_000061.1 | ||
| NM_024344.1 | NP_077320.1 | |||
| NM_173087.1 | NP_775110.1 | |||
| AF127764.1 | AAD28253.1 | |||
| AF127765.3 | AAD28254.3 | |||
| AK308103.1 | ||||
| BC146649.1 | ||||
| BC146672.1 | ||||
| EU791850.1 | ||||
| EU791851.1 | ||||
| X85030.1 | CAA59403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832986 | Chr.15:42295627 - 42461466 on Build GRCh38 | Loss |
|
| nsv832987 | Chr.15:42335249 - 42500150 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map