Assay Details
Target Gene Details
Entrez Gene ID: | 64799 |
Gene Name: | IQ motif containing H |
Gene Aliases: |
NYDSP5 |
Location: |
Chr.15:67254772-67501804 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827372 | Chr.15:67288578 - 67290795 on Build GRCh38 | Loss |
|
| esv3636778 | Chr.15:67287529 - 67290752 on Build GRCh38 | Loss |
|
| esv2666605 | Chr.15:67287523 - 67290748 on Build GRCh38 | Deletion |
|

Genomic Map